missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Swann-Morton™ Stainless Steel Surgical Scalpel Blades
£20.10 - £53.75
Specifications
Blade Material | Stainless Steel |
---|---|
Product Type | Surgical Blade |
Sterility | Sterile |
Quantity | 10/Pk. |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
12911645
|
Swann-Morton™
0324 |
# 19 |
£49.95
Pack of 100 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
12921645
|
Swann-Morton™
0306 |
# 20 |
£20.45
Pack of 100 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
12951645
|
Swann-Morton™
0309 |
# 22 |
£53.25
Pack of 100 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
12981645
|
Swann-Morton™
0312 |
# 25 |
£20.10
Pack of 100 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
12901655
|
Swann-Morton™
0313 |
26 |
£53.75
Pack of 100 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Specifications
Stainless Steel | |
Surgical Blade | |
Sterile | |
10/Pk. |