Learn More
Grant Instruments™ Stainless Steel Tank and Bridge Plate for Baths
Combine with any Grant Heating Circulator or Bath. Grant Instruments™ Stainless Steel Tank and Bridge Plate for Baths has a temperature range of –15 to 200°C.
Brand: Grant Instruments™ ST38
This item is not returnable.
View return policy
Description
Combine with any Grant Heating Circulator or Bath. Grant Instruments™ Stainless Steel Tank and Bridge Plate for Baths has a temperature range of –15 to 200°C.
- High efficiency stainless steel construction
- Templerature range: -15 to 200°C
- Includes integral raised feet for ease of carrying, repositioning, and retort access
Specifications
38 L | |
15°C to 200°C | |
730 mm |
Optima range immersion thermostats - T100, TC120, TXF150 or TXF200 | |
Stainless Steel |
Product Content Correction
Product Title
Grant Instruments™ Stainless Steel Tank and Bridge Plate for Baths > Capacity: 38L
Spot an opportunity for improvement?Share a Content Correction