Learn More
Hanna Instruments™ Stainless Steel Temperature Probes
AISI 316 stainless steel with wide measurement range that allows for temperature compensated measurements on compatible Hanna meters.
Brand:  Hanna Instruments™ HI7662
Product Code. 11587889
  • £248.00 / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless steel temperature probe with a 1 m cable length that allows for temperature compensated measurements on compatible Hanna meters
  • AISI 316 stainless steel
  • NTC thermistor
  • Wide measurement range
Specifications
Specifications
Temperature Probe
Stainless Steel