Learn More
Fisherbrand™ Stainless-Steel Test Sieve Stacks
Use these sieve stacks for research and development, quality control of raw materials, interim and finished products as well as in production monitoring. Fisherbrand™ Stainless-Steel Test Sieve Stacks are designed for use with AS 200 control vibratory sieve shakers.
Brand: Fisherbrand™ 60.131.000999
This item is not returnable.
View return policy
Description
- Sieve stack consisting of 8 test sieves (ISO 3310-1)
- 200mm diameter, 50mm height (45μm, 63μm, 125μm, 250μm, 500μm, 1mm, 2mm, 4mm)
- Includes collecting pan
Specifications
1 stack of 8 sieves | |
200 mm | |
Stainless Steel | |
Test Sieve Stack | |
Certificate of compliance with ISO 3310/1 or NFX 11-504, 8 Sieves (45μm, 63μm, 125μm, 250μm, 500μm, 1mm, 2mm, 4mm) and collecting pan |
For separation, fractioning, particle size determination | |
Stainless Steel | |
45μm, 63μm, 125μm, 250μm, 500μm, 1mm, 2mm, 4mm | |
ISO 3310 and ASTM E 11 | |
50mm |