missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Stainless-Steel Test Sieves, 100mm Dia., in Micrometer Pore Sizes with ISO 3310/1

For use with Vibratory Sieve Shaker AS 200 control

Brand:  RETSCH 60.106.000112

Code : NEW

Additional Details : Weight : 0.15000kg

 View more versions of this product

Product Code. 10449851

  • £89.00 / Each
Estimated Shipment: 14-10-2024
to see stock.
Add to basket

Description

Description

  • ISO 3310/1
  • Available in a wide range of pore sizes
Specifications

Specifications

Stainless Steel
Stainless Steel
1 Each
Test Sieve
Shaker AS 200
40mm
For separation, fractioning, particle size determination
100 mm
112μm
ISO 3310/1
Certificate indicating Compliance
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers