missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Stainless-Steel Test Sieves, 315 mm Dia., in Pore Size Greater than 10 mm

£255.00 - £275.40

Specifications

Quantity 1 Each
Type Test Sieve
Certifications/Compliance ISO 3310 and ASTM E 11, DIN 3310/2
For Use With (Application) For screening aggregates, separation, fractioning, particle size determination
Diameter (Metric) Outer 315 mm
View More Specs

Products 2
Product Code Brand Pore Size Price Quantity & Availability  
Product Code Brand Pore Size Price Quantity & Availability  
11732986
View Documents
RETSCH
60.172.014000
14 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11743006
View Documents
RETSCH
60.172.090000
90 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

1 Each
ISO 3310 and ASTM E 11, DIN 3310/2
315 mm
Stainless Steel
Test Sieve
For screening aggregates, separation, fractioning, particle size determination
Stainless Steel
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.