Learn More
RETSCH Stainless-Steel Test Sieves, 450mm Dia., in Micrometer Pore Sizes with ASTM E 11
£497.00 - £955.00
Specifications
Diameter (Metric) Outer | 450 mm |
---|---|
Frame Material | Stainless Steel |
Mesh Material | Stainless Steel |
For Use With (Application) | For separation, fractioning, particle size determination |
Quantity | 1 Each |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
11934702
|
RETSCH
60.169.000300 |
300μm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11994692
|
RETSCH
60.169.000150 |
150μm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11984692
|
RETSCH
60.169.000125 |
125μm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11974692
|
RETSCH
60.169.000106 |
106μm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11904702
|
RETSCH
60.169.000180 |
180μm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11914702
|
RETSCH
60.169.000212 |
212μm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11924702
|
RETSCH
60.169.000250 |
250μm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11914692
|
RETSCH
60.169.000038 |
38μm |
£955.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11944702
|
RETSCH
60.169.000355 |
355μm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11954692
|
RETSCH
60.169.000063 |
63μm |
£590.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11974702
|
RETSCH
60.169.000600 |
600μm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11954702
|
RETSCH
60.169.000425 |
425μm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11964702
|
RETSCH
60.169.000500 |
500μm |
£519.00
Each |
Estimated Shipment: 09-09-2024
Log in to see stock. |
|||||
11934692
|
RETSCH
60.169.000053 |
53μm |
£660.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11924692
|
RETSCH
60.169.000045 |
45μm |
£720.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11984702
|
RETSCH
60.169.000710 |
710μm |
£497.00
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
11944692
|
RETSCH
60.169.000075 |
75μm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11994702
|
RETSCH
60.169.000850 |
850μm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11964692
|
RETSCH
60.169.000090 |
90μm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
Specifications
450 mm | |
Stainless Steel | |
1 Each | |
ASTM E 11 | |
Certificate indicating Compliance |
Stainless Steel | |
For separation, fractioning, particle size determination | |
Test Sieve | |
Vibratory Shaker AS 450 | |
100mm |