Learn More
RETSCH Stainless-Steel Test Sieves, 450mm Dia., in Millimeter Pore Sizes with ASTM E 11
£497.00 - £635.00
Specifications
Diameter (Metric) Outer | 450 mm |
---|---|
Frame Material | Stainless Steel |
Mesh Material | Stainless Steel |
For Use With (Application) | For separation, fractioning, particle size determination |
Quantity | 1 Each |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
11944712
|
RETSCH
60.169.001700 |
1.7mm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11914712
|
RETSCH
60.169.001180 |
1.18mm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11924742
|
RETSCH
60.169.106000 |
106mm |
£635.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11914742
|
RETSCH
60.169.100000 |
100mm |
£635.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11954722
|
RETSCH
60.169.011200 |
11.2mm |
£620.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11924712
|
RETSCH
60.169.001400 |
1.4mm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11964722
|
RETSCH
60.169.012500 |
12.5mm |
£620.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11934742
|
RETSCH
60.169.125000 |
125mm |
£635.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11984722
|
RETSCH
60.169.016000 |
16mm |
£620.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11994722
|
RETSCH
60.169.019000 |
19mm |
£620.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11974722
|
RETSCH
60.169.013200 |
13.2mm |
£620.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11974712
|
RETSCH
60.169.002800 |
2.8mm |
£575.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11904712
|
RETSCH
60.169.001000 |
1mm |
£497.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
11954712
|
RETSCH
60.169.002360 |
2.36mm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11904732
|
RETSCH
60.169.022400 |
22.4mm |
£620.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11914732
|
RETSCH
60.169.025000 |
25mm |
£620.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11924732
|
RETSCH
60.169.026500 |
26.5mm |
£635.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11934712
|
RETSCH
60.169.002000 |
2mm |
£497.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11964712
|
RETSCH
60.169.003350 |
3.35mm |
£575.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11934732
|
RETSCH
60.169.031500 |
31.5mm |
£635.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11954732
|
RETSCH
60.169.037500 |
37.5mm |
£635.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11984712
|
RETSCH
60.169.004750 |
4.75mm |
£575.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11944732
|
RETSCH
60.169.045000 |
45mm |
£635.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11994712
|
RETSCH
60.169.004000 |
4mm |
£575.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11904722
|
RETSCH
60.169.005600 |
5.6mm |
£575.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11974732
|
RETSCH
60.169.053000 |
53mm |
£635.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11914722
|
RETSCH
60.169.006300 |
6.3mm |
£575.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11964732
|
RETSCH
60.169.050000 |
50mm |
£635.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11924722
|
RETSCH
60.169.006700 |
6.7mm |
£575.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11984732
|
RETSCH
60.169.063000 |
63mm |
£635.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11994732
|
RETSCH
60.169.075000 |
75mm |
£635.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11944722
|
RETSCH
60.169.008000 |
8mm |
£575.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11934722
|
RETSCH
60.169.009500 |
9.5mm |
£620.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11904742
|
RETSCH
60.169.090000 |
90mm |
£635.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
Specifications
450 mm | |
Stainless Steel | |
1 Each | |
ASTM E 11 | |
Certificate indicating Compliance |
Stainless Steel | |
For separation, fractioning, particle size determination | |
Test Sieve | |
Vibratory Shaker AS 450 | |
100mm |