missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Scaffolding Tubes with End Plugs

Provides for sturdy scaffolding construction in the laboratory. Fisherbrand™ Scaffolding Tubes with End Plugs are manufactured from durable stainless steel.

£14.85 - £51.10

Specifications

Material Stainless Steel
Product Type Tube
View More Specs

Products 3
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
11592662
View Documents Promotion Details
Fisherbrand™
2121/13
1000 mm
£26.50
Each
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
 
11502672
View Documents Promotion Details
Fisherbrand™
2121/43
1500 mm
£51.10
Each
Estimated Shipment: 29-11-2024
to see stock.
Add to basket
 
11582662
View Documents Promotion Details
Fisherbrand™
2121/33
250 mm
£14.85
Each
Estimated Shipment: 27-12-2024
to see stock.
Add to basket
 
Description

Description

  • Approx. 13mm diameter
  • Available in a range of lengths
  • Fits with Fisherbrand scaffolding connectors and foot adapters
Specifications

Specifications

Stainless Steel
Tube
Videos
SDS
Documents

Documents

Product Certifications