Learn More
Azpack™ Unex™ Tubing Clips
Product Code. 11328403
Click to view available options
Diameter (Metric):
10 to 14 mm
13 to 17 mm
14 to 19 mm
18 to 22 mm
21 to 25 mm
26 to 30 mm
28 to 33 mm
8 to 10 mm
Unit Size:
Pack of 5
Product Code. 11328403
Brand: Azpack™ 11328403
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Twist nut to constrict and release tubing. Azpack™ Stainless Steel Tubing Clips are designed to distribute and apply pressure evenly.
  • Stainless steel
  • Double-banded ring for even pressure
  • Nut and bolt included to apply proper compression
Specifications
Type Tubing Clamp
Material Stainless Steel (H10 304, A2)
Includes Tightening bolt
Diameter (Metric) 18 to 22 mm
Jaw Type Double-banded Ring