missing translation for 'onlineSavingsMsg'
Learn More

Fowler Vernier Caliper, Stainless Steel

Use this Stainless Steel Vernier Caliper to measure inside, outside, and depth. Features beveled edges for measuring screws.

Brand:  Fowler 09921-21

Product Code. 11774039

  • £111.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Features beveled edges for measuring screws.
Specifications

Specifications

Vernier Caliper
Vernier Caliper, Stainless Steel; 0 to 6 in./0 to 150 mm
0 to 150 mm (0 to 6 in.)
±0.001 in.
Stainless Steel
0.001 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fowler Vernier Caliper, Stainless Steel > Stainless Steel Vernier Caliper; Range: 0 to 150 mm (0 to 6 in.)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.