missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel wire, 0.25mm (0.01in) dia, Type 304, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 040945.BZ

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11365267

  • £40.00 / 150cm
In Stock
to see stock.

Description

Description

Stainless steel wire is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Stainless Steel wire, Type 304
Fe:Cr:Ni; 70:19:11 wt%
150 cm
Odorless
0.25mm (0.01 in.) dia.
MFCD00148499
Insoluble in water.
Wire
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only