missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Standard 3-Prong Extension Clamp, dual adjustable, medium, nickel-plated zinc

Designed to securely hold a large variety of glassware and apparatus. Cole-Parmer™ Standard 3-Prong Extension Clamps are ideal for irregularly shaped objects.

Brand:  Cole-Parmer™ 08021-34

Product Code. 11795429

  • £62.30 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

One prong or “finger” is adjusted using a wing nut, while the other prong remains stationary or for a more versatile clamp choose the dual adjustment style.
  • Utilizing thumbscrews, both prongs are easily adjusted over a wide range
  • Available in stainless steel or nickel plated zinc
  • Supplied with non-slip vinyl sleeves
  • Includes fiberglass covers temperatures above 100°C
  • Available in small, medium and large sizes
  • An additional holder must be purchased in order to attach clamps to lab-frames, support stands or other apparatus
Specifications

Specifications

3-Prong Extension Clamp
9.02 in.
Medium
0 to 2.72 in.
Nickel-plated Zinc
22.9 cm
0 to 69 mm
Dual Adjustment
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers