missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Standard Buffer Solution, SI Analytics™
SI Analytics™ Standard Buffer Solution pH 6.87 resists changes in pH when small quantities of an acid are added to it.
Specifications
Boiling Point | 100°C |
---|---|
Color | Colorless |
Density | 1g/cm3 |
pH | 6.87 |
Physical Form | Liquid |
Formulation | Water, disodium hydrogen phospate, potassium dihydrogen phosphate |
Odor | Odorless |
Packaging | FIOLAX™ Ampules |
Sterility | Sterile |
Vapor Pressure | 23hPa |
Description
- Includes manufacturer certificate
- Mixture, consisting of the following components: Water, disodium hydrogen phospate, potassium dihydrogen phosphate
- The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
Specifications
100°C | |
1g/cm3 | |
Liquid | |
Odorless | |
Sterile | |
Fully miscible in water |
Colorless | |
6.87 | |
Water, disodium hydrogen phospate, potassium dihydrogen phosphate | |
FIOLAX™ Ampules | |
23hPa |