missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Standard Buffer Solution pH 7, SI Analytics™
Click to view available options
Quantity:
60 Ampules
Description
- Includes manufacturer certificate
- Mixture, consisting of the following components: Water, disodium hydrogenphosphate, citric acid
- The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
- Absorb with liquid-binding material (sand, diatomite, acid binders, universal binders, sawdust)
Specifications
Boiling Point | 101°C |
Color | Colorless |
Density | 1.01g/cm3 |
pH | 7 |
Physical Form | Liquid |
Quantity | 60 Ampules |
Formulation | Water, disodium hydrogenphosphate, citric acid |
Odor | Odroless |
Packaging | FIOLAX™ Ampules |
Sterility | Sterile |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'