Learn More
Standard Color Solution YG (Yellowish Green) According to Chinese Pharmacopoeia (ChP), Reagecon
Chinese pharmacopeia standard color reference ampule.
Brand: Reagecon CPSRSYG01
Code : NEW
Additional Details : CAS Number : 7758-98-7 Weight : 0.10000kg
Description
- Standard yellowish green color reference
- Packaged in an amber glass Winchester bottle
Specifications
Amber Glass Winchester Bottle | |
Liquid |
Safety and Handling
- CHINESE PHARMACOPOEIA STANDARD SOLUTION GY (Yellowish Green)
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
Hazard Statement
- H350i-May cause cancer by inhalation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : copper sulphate, cobalt dichloride