Learn More
Kinematica Standard Dispersing Aggregate for PT3100 D
Uses two rows of teeth, is suitable for a large spectrum of laboratory applications, and is reliable and efficient.
Brand: Kinematica 11030298
Code : NEW
Additional Details : Weight : 0.40000kg
Description
- Selection and applications:
- All standard dispersing tasks
- Grinding of animal and human tissue samples in a variety of fluids and volumes
- Sample preparation for the extraction and dissolution of organic materials
- Sample preparation for medical diagnostic procedures as well as DNA analysis
- Extractions of systems / substances from vegetable-derived samples
- Disruption of animal and human cells
- Deagglomeration of united cell structures
- Intensive mixing
Specifications
Standard Dispersing Aggregate | |
0.3 to 10 mL | |
101 mm | |
1 Ea. |
8 m/s | |
7 mm | |
PT-DA 07 / 2EC-F101 |