Learn More
Welch™ Standard-Duty Vacuum Pumps
Compact, economical WOB-L™ pumps for aqueous applications
£417.96
Specifications
Voltage 115 V
View More Specs
Alert:
May be used with buffer solutions but are not recommended for use with acids, bases or organic vapors.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Products 1
Product Code Brand Model Flow Rate Price Quantity & Availability  
Product Code Brand Model Flow Rate Price Quantity & Availability  
15520741
View Documents
Welch™
U2511B-01
2511 11 L/min.
£417.96
Each
Description
Description
Compact, economical WOB-L™ pumps for aqueous applications
  • Ideal for filtration, vacuum ovens, aspiration and chamber roughing
  • Protect against accidental ingestion
  • Wetted parts resist corrosion from moisture
Order Info
For 115V 60Hz. All pumps weigh less than 25 lb. (11.4kg).
Specifications
Specifications
115 V