Learn More
Thermo Scientific™ Standard Fluid Transfer Assembly Extension Set
Simplify process connectivity and enable greater efficiency for most upstream and downstream process operations.
£131.35 - £790.46
Specifications
Content And Storage | Approved for storage at ambient conditions (15°C to 30°C). |
---|
Product Code | Brand | Tubing Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Style | Price | Quantity & Availability | |||||
17850677
|
Thermo Scientific™
SH31083.03 |
12.7mm (1/2 in) ID, 3.2mm (1/8 in) wall, C-Flex 374, 5m (16.4 ft) |
£790.46
Pack of 10 |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17830617
|
Thermo Scientific™
SH31079.02 |
3.2mm (1/8 in) ID, 1.6mm (1/16 in) wall, C-Flex 374, 2.5m (8.2 ft) |
£430.98
Pack of 10 |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17820617
|
Thermo Scientific™
SH31079.03 |
3.2mm (1/8 in) ID, 1.6mm (1/16 in) wall, C-Flex 374, 5m (16.4 ft) |
£390.03
Pack of 10 |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17850617
|
Thermo Scientific™
SH31080.01 |
6.35mm (1/4 in) ID, 1.6mm (1/16 in) wall, C-Flex 374, 1m (3.25 ft) |
£176.60
Pack of 10 |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17840617
|
Thermo Scientific™
SH31080.02 |
6.35mm (1/4 in) ID, 1.6mm (1/16 in) wall, C-Flex 374, 2.5m (8.2 ft) |
£131.35
Pack of 10 |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17830677
|
Thermo Scientific™
SH31080.03 |
6.35mm (1/4 in) ID, 1.6mm (1/16 in) wall, C-Flex 374, 5m (16.4 ft) |
£531.86
Pack of 10 |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17810677
|
Thermo Scientific™
SH31081.01 |
6.35mm (1/4 in) ID, 2.4mm (3/32 in) wall, C-Flex 374, 1m (3.25 ft) |
£366.48
Pack of 10 |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17860617
|
Thermo Scientific™
SH31081.02 |
6.35mm (1/4 in) ID, 2.4mm (3/32 in) wall, C-Flex 374, 2.5m (8.2 ft) |
£416.88
Pack of 10 |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17880617
|
Thermo Scientific™
SH31081.03 |
6.35mm (1/4 in) ID, 2.4mm (3/32 in) wall, C-Flex 374, 5m (16.4 ft) |
£502.93
Pack of 10 |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17870617
|
Thermo Scientific™
SH31082.01 |
9.52mm (3/8 in) ID, 3.2mm (1/8 in) wall, C-Flex 374, 1m (3.25 ft)) |
£382.63
Pack of 10 |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17840677
|
Thermo Scientific™
SH31082.02 |
9.52mm (3/8 in) ID, 3.2mm (1/8 in) wall, C-Flex 374, 2.5m (8.2 ft) |
£486.41
Pack of 10 |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17810617
|
Thermo Scientific™
SH31079.01 |
3.2mm (1/8 in) ID, 1.6mm (1/16 in) wall, C-Flex 374, 1m (3.25 ft) |
£698.60
Pack of 10 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17860677
|
Thermo Scientific™
SH31082.03 |
9.52mm (3/8 in) ID, 3.2mm (1/8 in) wall, C-Flex 374, 5m (16.4 ft) |
£581.05
Pack of 10 |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17810627
|
Thermo Scientific™
SH31083.01 |
12.7mm (1/2 in) ID, 3.2mm (1/8 in) wall, C-Flex 374, 1m (3.25 ft) |
£229.89
Pack of 10 |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17890617
|
Thermo Scientific™
SH31083.02 |
12.7mm (1/2 in) ID, 3.2mm (1/8 in) wall, C-Flex 374, 2.5m (8.2 ft) |
£271.63
Pack of 10 |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Approved for storage at ambient conditions (15°C to 30°C). |