Learn More
Thermo Scientific™ Standard Fluid Transfer Assembly Y Set
Shop All Thermo Scientific ProductsDescription
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Spécification
Spécification
| Dimensions | 3/8 in. ID x 1/8 in. wall |
| Content And Storage | Approved for storage at ambient conditions (15°C to 30°C). |
| Tubing Style | 3 x 9.52mm (3/8 in) ID C-Flex 374 |
| Tubing Size | 2 x 0.51m (20 in) to 0.51m (20 in) |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.