missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Standard Ground Joint Funnel
Half-round body
Brand: BRAND™ 94915
26.53 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
80 mm | |
Stem for NS 29/32 | |
Standard Ground Joint Funnel |
Polypropylene | |
5 |