Learn More
Koettermann™ Standard Hanging Cabinet, 1200x630x366mm
Click to view available options
Locking Mechanism:
No
Yes
Unit Size:
Each
Description
- Wall-mounted cabinet
- Two doors
- One shelf
- Side panels
Specifications
Specifications
| Product Type | Wall-mounted Cabinet |
| Dimensions (D x W x H) | 366 x 1200 x 630 mm |
| Locking Mechanism | No |
| No. of Doors | 2 |
| No. of Shelves | 1 |