Learn More
METTLER TOLEDO Standard MA Analytical Balance
Product Code. 18062364 Shop All Mettler Toledo Products
Click to view available options
Capacity:
120 g
220 g
52 g
62 g, 152 g
92 g
Model:
MA104
MA155DU
MA204
MA54
MA55
MA95
Readability:
0.01 mg
0.01 mg, 0.1 mg
0.1 mg
Unit Size:
Each
Product Code. 18062364
Brand: METTLER TOLEDO 30706701
METTLER TOLEDO Standard MA Analytical Balances are highly accurate, efficient, simple to operate and designed with robust construction for proven long lasting performance.
The LCD hybrid touchscreen is simple to operate and will increase efficiency of your daily tasks with 9 built-in applications and statistical data analysis. Built with a metal base, PBT top housing and overload protection to defend against rough handling in any weighing environment, ensuring quality performance for years to come.
MA Analytical Balances include:
  • Internal adjustment
  • LCD hybrid touchscreen with onscreen instructions
  • 9 built-in applications
  • Statistical data analysis
  • Front level indicator
  • Metal base, PBT top housing
  • Overload protection
  • 2 interfaces: USB-A, RS232
  • Bluetooth option
The MA Balance is the successor to the ME-T Balance. See how it ranks among our balance lines:
  • XPR Excellence Analytical Balances
  • MX Advanced Analytical Balances
  • MR Advanced Analytical Balances
  • MA Standard Analytical Balances
  • ME Standard Analytical Balances
  • LA Standard Analytical Balances
TRUSTED_SUSTAINABILITY
Specifications
Connection Type USB-A, RS232
Capacity 92 g
Display Type Touchscreen
Readability 0.01 mg
Material Metal, PBT
Model MA95
Stabilization Time 4 sec.
Type Analytical Balance
Electrical Requirements 100/240 V, 50/60 Hz
Legal for Trade No