Learn More
Thermo Scientific™ Standard Plate Count Agar (APHA) (Dehydrated)
Determine the plate counts of microorganisms in food, dairy samples, waste water and other materials of sanitary importance with this medium.
£279.00 - £1225.00
Specifications
Certifications/Compliance APHA, AOAC, PHLS, ISO 11133:2014
Form Powder
Product Type Agar
Test Type Standard Plate Count for Microorganism
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10679554
View Documents Promotion Details
Thermo Scientific™
CM0463B
500 g
£279.00
500g
In Stock
to see stock.
Add to basket
13205719
View Documents
Thermo Scientific™
CM0463T
5 kg
£1225.00
5kg
Estimated Shipment: 07-03-2025
to see stock.
Add to basket
Description
Description
Determine the plate counts of microorganisms in food, dairy samples, waste water and other materials of sanitary importance with this medium.
  • Meets formulation of APHA, AOAC and PHLS
  • Quality control of Standard Plate Count Agar (APHA) includes testing in accordance with ISO 11133:2014
Specifications
Specifications
APHA, AOAC, PHLS, ISO 11133:2014
Powder
Agar
Standard Plate Count for Microorganism
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Standard Plate Count Agar (APHA) (Dehydrated)