missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Standard Rack for Microtiterplates, for Upright Freezers, 65 mm Shelf Height
Stainlesss Steel Standard Rack for use in upright freezers is designed to hold microtiterplates.
Brand: Tenak™ TE27604
This item is not returnable.
View return policy
Description
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of stainless steel
Specifications
4 x 7 | |
Stainless Steel | |
630 x 135 x 266 mm |
Microtiterplates (28) | |
Upright Freezers | |
65 mm |