Learn More
Spectrum™ Labs Standard RC Dialysis Disc
Product Code. p-8011024
Click to view available options
Diameter (Metric):
33 mm
100 mm
Unit Size:
Pack of 50
Product Code. 12727526
Brand: Spectrum™ Labs 132482
For standard grade RC dialysis tubing
For use with Spectrum Equilibrium Dialyzer and Spectra/Por™ MacroDialyzer systems.
  • Regenerated cellulose (RC) precut membrane discs
  • Supplied dry with a glycerol as a humectant
Specifications
Product Type Standard RC Dialysis Disc
Diameter (Metric) 100 mm
Quantity 50 Pk.
For Use With (Application) For standard grade RC dialysis tubing