missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Standard RC Dialysis Tubing
Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes
Brand: Spectrum™ 132703
This item is not returnable.
View return policy
Description
Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes
- Membrane grade
- Membrane type: RC
- Molecular weight cut-off: 12 to 14K
Specifications
Spectra/Por 4 Dialysis Tubing | |
Regenerated Cellulose | |
0.8 in. | |
30 m | |
39.2°F to 249.8°F | |
100 Pack |
12 to 14 kDa | |
20.4 mm | |
100 ft. | |
Contain glycerin as a humectant | |
4°C to 121°C | |
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction