missing translation for 'onlineSavingsMsg'
Learn More

Standard Color Solution BY, Brown/Yellow, EP Grade, Reagecon™

Used to calibrate, control, qualify and validate color measurement instruments. Standard Color Solution BY, Brown/Yellow, EP Grade, Reagecon™ is a ready-to-use colorimetric solution produced in accordance with Eu. Ph. Chap 2 from a precise combination of Primary Color Solutions.

Brand:  Reagecon EP704

Additional Details : Weight : 0.13925kg Transport : UN number : 1760 Chem class : 8 Pack group : III


Product Code. 11894242

  • £210.00 / 125mL
Estimated Shipment: 26-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ready-to-use colorimetric standard presented in a high-quality, tamper-evident bottle. EP Standard Color Solutions are prepared from the EP Primary Color Solutions (yellow, red and blue) and can be used to prepare EP Reference Color Solutions.
  • Manufactured according to European Pharmacopoeia Chapter 2 guidelines
  • Balance Certificate of Calibration issued in accordance with ISO 17025
  • Prepared gravimetrically on a weight/weight basis using OIML traceable weights
  • Concentration verified using a high-performance spectrophotometer calibrated using ISO Guide 34 standards
  • Color Standard BY = 2.4 Yellow + 1.0 Red + 0.4 Blue + 6.2 Hydrochloric Acid (10g HCl/L)
Specifications

Specifications

European Pharmacopoeia Standard Color Solution BY
125 mL
Brown/Yellow
Eu. Ph. Chapter 2
Liquid
Tamper evident
7705-08-0,7646-79-9,7758-98-7,7647-01-0
Yes
Iron chloride, Cobalt chloride, Copper sulfate, Hydrochloric acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • STANDARD SOLUTION BY (BROWN / YELLOW)

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1B
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H350i-May cause cancer by inhalation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • EUH208-Contains . May produce an allergic reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.

Supplemental information
  • MIXTURE LIST-Contains : copper sulphate, hydrochloric acid 37%,iron trichloride, cobalt dichloride

SDS
Documents

Documents

Certificates
Special Offers

Special Offers