missing translation for 'onlineSavingsMsg'
Learn More

Standard Color Solution R, Red, EP Grade, Reagecon™
BIOPHARMA

Used to calibrate, control, qualify and validate color measurement instruments. Standard Color Solution R, Red, EP Grade, Reagecon™ is a ready-to-use colorimetric solution produced in accordance with Eu. Ph. Chap 2 from a precise combination of Primary Color Solutions.

Brand:  Reagecon EP707


Product Code. 11804252

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ready-to-use colorimetric standard presented in a high-quality, tamper-evident bottle. EP Standard Color Solutions are prepared from the EP Primary Color Solutions (yellow, red and blue) and can be used to prepare EP Reference Color Solutions.
  • Manufactured according to European Pharmacopoeia Chapter 2 guidelines
  • Balance Certificate of Calibration issued in accordance with ISO 17025
  • Prepared gravimetrically on a weight/weight basis using OIML traceable weights
  • Concentration verified using a high-performance spectrophotometer calibrated using ISO Guide 34 standards
  • Color Standard R = 1.0 Yellow + 2.0 Red + 7.0 Hydrochloric Acid (10g HCl/L) 
Specifications

Specifications

European Pharmacopoeia Standard Color Solution R
100 mL
Red
Eu. Ph. Chapter 2
Liquid
Tamper evident bottle
7705-08-0, 7646-79-9, 7647-01-0
Yes
Iron chloride, Cobalt chloride, Hydrochloric acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • STANDARD SOLUTION R (RED) EUROPEAN PHARMACOPOEIA

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin sensitiser Category 1

Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H411-Toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid 37%, cobalt dichloride

SDS
Documents

Documents

Certificates
Special Offers

Special Offers