Learn More
Fisherbrand™ Standard Vacuum Pump
Use these pumps for a wide range of laboratory applications and continuous running. Fisherbrand™ Standard Vacuum Pumps provide a high water vapor tolerance and are ideal features for both research and routine applications.
Brand:  Fisherbrand™ 400170-15
Product Code. 11959078
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Compact design, low weight and easy to use
  • Quiet running and excellent ultimate vacuum
  • Active corrosion protection
  • Easy maintenance
  • Supplied with clamps and seals for the inlet and outlet
  • Each pump comes with anti-suck back valve, gas ballast valve, oil charging, centering ring, clamping ring and motor overload protection
Specifications
Specifications
235 mm
16 L/min.
145 mm
Vacuum Pump
230 V
PTFE
50 Hz
195 mm
IP54
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products