Learn More
3M™ Standard Visor
Replacement visor for the 3M™ Reusable Full Face Mask FF-600 Series.
Brand: 3M™ 7100266117
184.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.82000kg
Description
- Replacement visor for the 3M™ Reusable Full Face Mask FF-600 Series
- Can be fitted by the user or a service center if the original visor becomes lost or damaged.
Specifications
Replacement Visor | |
3M™ Reusable Full Face Mask FF-600 Series | |
5/Cs. |
Visor | |
FF-600F-925 |