Learn More
Cole-Parmer™ Stack Blocks for Roller Shakers
Stack up to 3 shakers for bench saving space with these Stack Blocks for Roller Shakers.
Brand: Cole-Parmer™ 04653-09
Code : 60
Additional Details : Weight : 0.01000kg
Description
- Kit contains four blocks.
- A single kit will stack two units — order two kits to stack three tube roller shakers.
Specifications
Stacking System | |
4 Staple Blocks |