missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch, for analysis, soluble, Thermo Scientific Chemicals
£38.89 - £362.81
Chemical Identifiers
CAS | 9005-84-9 |
---|---|
Molecular Formula | C12H22O11 |
Molecular Weight (g/mol) | 342.297 |
MDL Number | MFCD00082026 |
InChI Key | GUBGYTABKSRVRQ-ASMJPISFSA-N |
Synonym | alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose |
PubChem CID | 439341 |
ChEBI | CHEBI:18167 |
IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol |
SMILES | C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10572581
|
Thermo Scientific Acros
177132500 |
250 g | Plastic bottle |
£38.89
250g |
In Stock
Log in to see stock. |
||||
10337980
|
Thermo Scientific Acros
177130010 |
1 kg | Plastic bottle |
£103.52
1kg |
In Stock
Log in to see stock. |
||||
10424605
|
Thermo Scientific Acros
177130050 |
5 kg | Plastic drum |
£362.81
5kg |
Estimated Shipment: 17-12-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9005-84-9 | |
342.297 | |
GUBGYTABKSRVRQ-ASMJPISFSA-N | |
439341 | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol |
C12H22O11 | |
MFCD00082026 | |
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
CHEBI:18167 | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O |
Specifications
White | |
9005-84-9 | |
256.0°C to 258.0°C | |
MFCD00082026 | |
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
GUBGYTABKSRVRQ-ASMJPISFSA-N | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol | |
439341 | |
Analytical | |
Authentic | |
passes |
5 to 7.5 (2% soln. at 25°C) | |
Amorphous Powder | |
C12H22O11 | |
15, 8926 | |
Solubility in water: 50g/L (90°C). | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O | |
342.297 | |
CHEBI:18167 | |
positive | |
10% max. | |
Starch, Soluble, For analysis |
RUO – Research Use Only