missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch, Certified AR for Analysis, Soluble, Fisher Chemical™
C12H22O11, CAS Number-9005-25-8, starch, soluble, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose, glcalpha1-4glca, maltose, unii-15sug9ad26, starch solution, alpha-malt sugar, alpha-maltose, amylodextrin, glcalpha1-4glcalpha, 250g, CHEBI:18167, White, GUBGYTABKSRVRQ-ASMJPISFSA-N
£74.10 - £205.00
Chemical Identifiers
CAS | 9005-25-8 |
---|---|
Molecular Formula | C12H22O11 |
Molecular Weight (g/mol) | 342.30 |
MDL Number | MFCD00082026,MFCD00132834 |
InChI Key | GUBGYTABKSRVRQ-UHFFFAOYNA-N |
Synonym | alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose |
PubChem CID | 439341 |
ChEBI | CHEBI:18167 |
IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol |
SMILES | OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10677612
|
Fisher Chemical
S/8000/50 |
250 g |
£74.10
250g |
In Stock
Log in to see stock. |
|||||
10427270
|
Fisher Chemical
S/8000/53 |
500 g |
£122.00
500g |
In Stock
Log in to see stock. |
|||||
10447460
|
Fisher Chemical
S/8000/60 |
1 kg |
£205.00
1kg |
In Stock
Log in to see stock. |
|||||
Description
C12H22O11, CAS Number-9005-25-8, starch, soluble, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose, glcalpha1-4glca, maltose, unii-15sug9ad26, starch solution, alpha-malt sugar, alpha-maltose, amylodextrin, glcalpha1-4glcalpha, 250g, CHEBI:18167, White, GUBGYTABKSRVRQ-ASMJPISFSA-N
Chemical Identifiers
9005-25-8 | |
342.30 | |
GUBGYTABKSRVRQ-UHFFFAOYNA-N | |
439341 | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol |
C12H22O11 | |
MFCD00082026,MFCD00132834 | |
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
CHEBI:18167 | |
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O |
Specifications
White | |
9005-25-8 | |
C12H22O11 | |
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O | |
342.30 | |
CHEBI:18167 |
5 to 7 | |
Powder Solid | |
MFCD00082026,MFCD00132834 | |
GUBGYTABKSRVRQ-UHFFFAOYNA-N | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol | |
439341 | |
Plastic powder jar |