Learn More
Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final Volume 1L, Thermo Scientific Chemicals
For Iodometric Titrations
£75.00 - £296.00
Specifications
Chemical Name or Material | Starch indicator solution |
---|---|
CAS | 9005-84-9, 7732-18-5 |
Physical Form | Liquid,For iodometric titrations |
Assay Percent Range | 100% |
MDL Number | MFCD00082026 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11389329
|
Thermo Scientific Alfa Aesar
088632.LQ |
1 U |
£75.00
1 unit |
In Stock
Log in to see stock. |
|||||
11399329
|
Thermo Scientific Alfa Aesar
088632.LS |
6 U |
£296.00
6 units |
In Stock
Log in to see stock. |
|||||
Description
Starch indicator 1% solution is used as an indicator for redox titrations. It is also used in the detection of end point in iodometric titration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Starch indicator solution | |
Liquid,For iodometric titrations | |
MFCD00082026 | |
Starch: 20%; Water: 80% |
9005-84-9, 7732-18-5 | |
100% | |
Miscible with water. | |
17mm Hg at 20°C |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only