missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations), Thermo Scientific™
For iodometric titrations
£23.80 - £144.00
Specifications
CAS | 9005-84-9 |
---|---|
Molecular Formula | C12H22O11 |
Molecular Weight (g/mol) | 342.297 |
MDL Number | MFCD00082026 |
InChI Key | GUBGYTABKSRVRQ-ASMJPISFSA-N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15434125
|
Thermo Scientific Alfa Aesar
047209.AP |
500mL |
£23.80
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15447475
|
Thermo Scientific Alfa Aesar
047209.K2 |
1L |
£29.20
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15457475
|
Thermo Scientific Alfa Aesar
047209.K7 |
4L |
£55.60
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15444125
|
Thermo Scientific Alfa Aesar
047209.M4 |
20L |
£144.00
20 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
9005-84-9 | |
342.297 | |
GUBGYTABKSRVRQ-ASMJPISFSA-N | |
439341 | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol |
C12H22O11 | |
MFCD00082026 | |
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
CHEBI:18167 | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O |
Safety and Handling
missing translation for 'einecsNumber' : 232-686-4
missing translation for 'rtecsNumber' : GM5090000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only