missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch Indicator Solution, Chem Lab™
£148.00
Specifications
Melting Point | 0°C |
---|---|
Solubility Information | Soluble in water |
Odor | Odorless |
Color | Colorless |
pH | ±7 |
Physical Form | Liquid |