missing translation for 'onlineSavingsMsg'
Learn More
Starch, Soluble, Reagent ACS, Thermo Scientific Chemicals Product Code.: 10361361

Starch, Soluble, Reagent ACS, Thermo Scientific Chemicals

Product Code. 10361361
2 kg, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10361361

Brand: Thermo Scientific Acros 424490020

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-84-9
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.297
MDL Number MFCD00082026
InChI Key GUBGYTABKSRVRQ-ASMJPISFSA-N
Synonym alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439341
ChEBI CHEBI:18167
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O

Specifications

Color White
pH 5.0 to 7.0 (2% soln. at 25°C)
Physical Form Powder
Melting Point 256.0°C to 258.0°C
Solubility Information Solubility in water: 50g/L (90°C).
Quantity 2 kg
Grade ACS Reagent
Infrared Spectrum Authentic
Packaging Plastic bottle
Residue after Ignition 0.4% max.
Sensitivity Passes Test
Chemical Name or Material Starch, Soluble
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0471792 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number424491000,424490020,424490100,424495000
Lot NumberA0468679 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number424491000,424490020,424490100,424495000
Lot NumberA0468678 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number424491000,424490020,424490100,424495000
Lot NumberA0467100 Certificate TypeCertificate of Analysis Date31/10/2024 Catalog Number424491000,424490020,424490100,424495000
Lot NumberA0467099 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog Number424491000,424490020,424490100,424495000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Starch, Soluble, Reagent ACS, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.