missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch, Soluble, Reagent ACS, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White |
pH | 5.0 to 7.0 (2% soln. at 25°C) |
Physical Form | Powder |
Melting Point | 256.0°C to 258.0°C |
Solubility Information | Solubility in water: 50g/L (90°C). |
Quantity | 10 kg |
Grade | ACS Reagent |
Infrared Spectrum | Authentic |
Packaging | Plastic drum |
Residue after Ignition | 0.4% max. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'