Learn More
Starch Solution, EP Grade, Reagecon™

Manufactured according to Eu. Ph. Chapter 4 (4.1.1) and prepared in an ISO 9001 facility. Starch Solution, EP Grade, Reagecon™ is a ready-to-use reagent with 1g soluble starch in a solution of water and mercuric iodide for use in manual and automated titrations, including redox titrations.
Brand: Reagecon 1085103
Description
Starch solution is often used an indicator reagent in redox titrations, where it turns blue in the presence of an oxidizing agent and colorless with exposure to a reducing agent.
This starch solution is prepared by triturating 1.0g soluble starch with 5mL water and then stirring the mixture into 100mL boiling water containing 10mg mercuric iodide.
- Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.1.1) guidelines
- Provided with a certificate of analysis and a label that shows the lot and expiration date
- Ready to use product eliminates the time and labor of preparing precise reagents and volumetric solutions
Specifications
Soluble starch | |
Undesignated | |
Liquid | |
1.0g soluble starch + 5mL water R, poured into 100mL boiling water + 10mg mercuric iodide | |
Tamper evident | |
342.30g/mol |
9005-84-9, 7774-29-0, 7732-18-5 | |
Yes | |
100 mL | |
C12H22O11 | |
Amylodextrin solution, Soluble starch solution | |
Eu. Ph. Chapter 4(4.1.1) |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1851325B1 | Certificate of Analysis | 20/03/2025 | 1085103 |
1851324M1 | Certificate of Analysis | 04/03/2025 | 1085103 |
1851324J1 | Certificate of Analysis | 17/12/2024 | 1085103 |
1851324F1 | Certificate of Analysis | 15/10/2024 | 1085103 |
1851324B1 | Certificate of Analysis | 19/06/2024 | 1085103 |
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
0Hours
14Minutes
28Seconds
21
Your input is important to us. Please complete this form to provide feedback related to the content on this product.