missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch Solution, Honeywell Fluka™
Indicator, 1% in H2O
Brand: Honeywell Fluka 319554-100ML
Code : NEW
Additional Details : Weight : 0.10050kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Starch Solution | |
1.005g/mL at 25°C | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
100 mL |