Learn More
Heidolph instruments StarFish Base Plate
Maintain good thermal contact with your heated magnetic plate stirrer. Heidolph™ StarFish Base Plate fits securely on the top plate and can be outfitted with optional safety handles.
Brand:  Heidolph instruments 505-81000-00
Product Code. 10545676
  • £663.00 / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Securely attaches to top plate.
  • Maintains good thermal contact.
  • Optional safety handles.
Specifications
Specifications
145 mm
Starfish Workstation, MR-Hei Magnetic Stirrers