Learn More
Heidolph™ StarFish Safety Handles
Pair of handles for safely lifting the base plate from the StarFish Multi-Experiment Workstation.
Brand: Heidolph™ 505-81001-00
Additional Details : Weight : 0.39200kg
Description
- Particularly relevant for chemical reactions that require heat supply
- Insulated handles can be screwed firmly to base plate for safe removal of system from hot magnetic stirrer plate
- Handles come as a pair
Specifications
StarFish Multi-Experiment Workstation | |
Lifting Base Plate | |
Insulated Handles |