missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences StarGate pLSG-IBA Vectors for Baculo
GREENER_CHOICE

Product Code. p-7147235 Shop All IBA Lifesciences Products
Click to view available options
Vector:
pLSG-IBA 102
pLSG-IBA 103
pLSG-IBA 104
pLSG-IBA 105
pLSG-IBA 123
pLSG-IBA 142
pLSG-IBA 143
pLSG-IBA 144
pLSG-IBA 145
pLSG-IBA 162
pLSG-IBA 164
pLSG-IBA 167
pLSG-IBA 168
pLSG-IBA 23
pLSG-IBA 25
pLSG-IBA 3
pLSG-IBA 33
pLSG-IBA 35
pLSG-IBA 43
pLSG-IBA 45
pLSG-IBA 5
pLSG-IBA 62
pLSG-IBA 63
pLSG-IBA 64
pLSG-IBA 65
pLSG-IBA wt1
pLSG-IBA wt2
Unit Size:
5µg
This item is not returnable. View return policy

Product Code. 15623828

Brand: IBA Lifesciences 54800001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Transfer vectors that allow for high-level expression in insect cells by introducing the gene of interest into the Polyhedrin gene locus of AcMNPV DNA by homologous recombination.

pLSG-IBA Baculo virus (insect cells) expression vectors

The pLSG-IBAwt1 vector carries the Polyhedrin promoter for high-level expression in insect cells.

  • Co-transfection with BacPAK6 linearized AcMNPV DNA (Clontech) or circular flashBAC modified AcMNPV DNA (Oxford Expression Technologies) allows the generation of recombinant baculovirus at very high efficiency through reconstitution of an essential gene (ORF 1629) and elimination of wild type virus to great extent.
  • Cloning and expression of recombinant proteins in insect cells
  • Polyhedrin promoter drives high-level expression of the insert
  • Ampicillin resistance and ColE1 origin of replication (pUC) support propagation in E. coli

Please note that cloning into pLSG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pLSG-IBA vectors with Esp3I, another option via a so-called Entry-Vector is possible.

Specifications

Format Liquid
For Use With (Equipment) StarGate™ System, Insect Host Cells
Inducing Agent BacPAK6 linearized A cmNPV DNA (Clontech) or circular flashBAC modified A cmNPV DNA (Oxford Expression Technologies)
Includes pUC ori, Ampicillin resistance
pH 8
Promoter Polyhedrin
Product Type Acceptor Vector for bacula (insect cell) expression
Storage Buffer 20 μL TE buffer: 10 mm Tris-HCl, 1 mm EDTA / 5 μg
Content And Storage 2°C to 8°C for frequent usage, -20°C for long-term storage
Protein Tag Untagged
Concentration 250 ng/μL
Quantity 5 μg
Vector pLSG-IBA wt1
Color Clear
For Use With (Application) Cloning Genes of Interest for Use in Functional Expression, Purification, Detection, Immobilization or Separation of Recombinant Proteins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.