Learn More
Elma™ Starter Kit
Click to view available options
:
Starter Kit; For Use With (Equipment): Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
Description
- Includes: 3 of each Elma Pin (0741 and 0772), 2 of each Elma Pin (0509, 0158, 0767, 0613 and 0614, 0021R and 0001R), 2 of each silicone holder side comb and bottom comb
- Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
- Ergonomic handles
- Autoclavable
- Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
- Square openings in the side walls for mounting silicone holders
Specifications
Specifications
| Type | Starter Set |
| For Use With (Equipment) | Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |