missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Starter Pak No. 1
Features a combination of yeast- and soy-based peptones.
Brand: Gibco™ 215366
This item is not returnable.
View return policy
Description
- Three of the products in this pack have been ultra-filtered to reduce endotoxin levels
- The yeast products add a mixture of peptides, amino acids, carbohydrates (simple and complex), nucleosides, and vitamins to any medium formulation
- Starter Pak No. 1 provides conveniently packaged, 100-g, non-GMP samples of these peptones that are best suited for use in human and animal health applications and contains:
- Difco TC Yeastolate UF and Bacto TC Yeastolate—these peptones are ideal for CHO-based applications of biotherapeutic monoclonal antibodies and recombinant proteins
- Difco Yeast Extract UF and Bacto Yeast Extract, Technical—these peptones support optimal growth of many microbial species for a variety of human and animal health vaccines
- Difco Phytone Supplement, UF—this enzymatic digest of soy is an excellent, nutritious source of carbohydrates and is used in mammalian cell culture. This peptone works well alone and when blended with yeast-based peptones
- Our Starter Paks provide conveniently packaged samples of the peptones most commonly used in mammalian cell culture and microbial fermentation bioproduction processes. Starter Paks are tailored for specific applications, including the production of monoclonal antibodies, recombinant proteins, and vaccines.

Specifications
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. | |
Powder | |
Human and animal health applications | |
Animal origin-free, yeast and soy |
Microbial | |
Dehydrated Culture Media | |
1 Ea. |
User Guide: Starter Pak No. 1
Version: B.0
File size: 105.5 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Starter Pak No. 1 > Quantity: 1 each
Spot an opportunity for improvement?Share a Content Correction