Learn More
Eppendorf Starter Set 100 μL 384-format: Eppendorf Xplorer™ Plus Move It™
Move It, the adjustable tip spacing pipette from Eppendorf, allows easy vessel format changes while saving up to 50 % time compared to single-channel pipettes. Its unique tubeless design allows autoclavability. Available as mechanical or electronic pipette with 4, 6, 8, or 12 channels.
Brand:  Eppendorf 4861000954
 View more versions of this product
Product Code. 30058129
  • £3476.85 / 1 set
Explore more special offers
Description
Description
  • 4-, 6-, 8- and 12-channel air-cushion pipettes with variable volume and adjustable tip spacing option
  • Available as mechanical (Research plus) and electronic (Xplorer plus) adjustable tip spacing multi-channel pipette
  • For plates (384-, 96-, 48-, 24- and 12-well), tubes, agarose gels and more (depending on pipette variant)
  • Tip spacing freely selectable from 4.5 to 33 mm, depending on pipette variant
  • Format limiter for quick switches backwards and forwards between source and destination formats
  • Manual adjustment knob for smooth vibration-free format switches and liquid transfers without dripping
  • Tubeless system for enhanced reliability and safety
  • Autoclavable for user and sample safety (Research plus fully, and Xplorer plus lower part only)
  • 360° rotatable pipette head for comfortable readability of display and fast identification of parameters
  • Optimally balanced pipette - reduces hand fatigue for stress-free operation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lot-Specific Certificate
Autoclavable
Single-use
Sterile
5 to 100 μL
1 Xplorer™ plus Move It™, adjustable cone distance, 8-channel, 5 to 100 μL, light yellow with 5x ep Dualfilter T.I.P.S.™, PCR clean and sterile, 5 to 100 μL, 53 mm, yellow, 19020 tips (50 racks x 384 tips)
2.1 in.
epT.I.P.S.™, epT.I.P.S.™ 384
Light Yellow
8
Manual Pipettes
Air-cushion System
19,020 Tips (50 Racks x 384 Tips)
53 mm