missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf Starter Set 100 μL 384-format: Eppendorf Xplorer™ Plus Multichannel

From blue sky research to clinical applications, Eppendorf Xplorer / Xplorer plus electronic pipettes provide simplicity, precision, and reproducibility. Available as single-, 8-, 12-, 16-, and 24-channel pipettes, as well as Move It™ adjustable tip spacing variant, for your challenging work.

Brand:  Eppendorf 4861000953

2455.81 GBP valid until 2024-12-06
Use promo code "21633" to get your promotional price.


 View more versions of this product

Product Code. 30058128

  • £3315.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Electronic air-cushion pipette reaching new levels of simplicity, precision and reproducibility
  • Intuitive handling via multi-function rocker with 'up is up' (aspiration) and 'down is down' (dispensing) logic and selection dial to access all pipetting mode
  • Self-explanatory color display without cluttered submenus and with ergonomic viewing angle for optimal readability in every position
  • Spring-loaded tip cone in all Xplorer and Xplorer plus pipettes up to 1,000 μL reduces tip attachment and ejection forces
  • Powerful rechargeable battery for up to 8 hours use & work can continue while charging via power socket
  • Pre-programmed and individual adjustment to external conditions (e.g. geographic height, liquids with high density, elongated tips)
  • Autoclavable lower parts for easy decontamination (all models, incl. Move It™)
  • 9 display languages: Chinese, Dutch, English, French, German, Italian, Japanese, Portuguese and Spanish
  • Xplorer plus only: Individual programming, fixed volume settings, password protection, additional applications, display of service intervals
  • ACT™ - labeled single-channel pipettes and epT.I.P.S.™ pipette tips help you make an informed choice when it comes to sustainability
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lot-Specific Certificate
Autoclavable
Single-use
Sterile
5 to 100 μL
Air-cushion System
19,020 Tips (50 Racks x 384 Tips)
epT.I.P.S.™, epT.I.P.S.™ 384
Light Yellow
24
Electronic Pipettes
Pipetting of aqueous liquids
1 Xplorer™ plus, 24-channel, variable, 5 to 100 μL, light yellow with 5x ep Dualfilter T.I.P.S.™, PCR clean and sterile, 5 to 100 μL, 53 mm, yellow, 19020 tips (50 racks x 384 tips)
0.0416666667
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers