missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ StartingBlock™ Blocking Buffer
StartingBlock Blocking Buffer is a single purified protein for fast blocking of western blots and ELISA assays.
£59.00 - £271.00
Specifications
Product Type | Blocking Buffer |
---|---|
Form | Liquid |
Product Line | StartingBlock™ |
Blocking Agent | Single purified protein |
Content And Storage | Upon receipt store at 4°C. |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
13414299
|
Thermo Scientific™
37578 |
StartingBlock (PBS) Blocking Buffer, 100 mL |
£59.00
100mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10108313
|
Thermo Scientific™
37538 |
StartingBlock (PBS) Blocking Buffer, 1 L |
£271.00
1 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13424299
|
Thermo Scientific™
37579 |
StartingBlock (TBS) Blocking Buffer, 100 mL |
£59.00
100mL |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10371804
|
Thermo Scientific™
37542 |
StartingBlock (TBS) Blocking Buffer, 1 L |
£269.00
1 L |
Estimated Shipment: 25-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10270404
|
Thermo Scientific™
37539 |
StartingBlock T20 (PBS) Blocking Buffer, 1 L |
£266.00
1 L |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10381804
|
Thermo Scientific™
37543 |
StartingBlock T20 (TBS) Blocking Buffer, 1 L |
£266.00
1 L |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
StartingBlock Blocking Buffer is a single purified protein for fast blocking of western blots and ELISA assays. It provides broad compatibility with a wide range of antibodies, antibody combinations, and other protein probing and assay systems. StartingBlock Blocking Buffer is available in PBS and TBS, with and without Tween 20.
Features of StartingBlock Blocking Buffer include:• Strip and re-probe without re-blocking—blots stay blocked even after stripping with Restore Stripping Buffer (Cat. No. 21059)
• Compatible with many detection systems—western blot, ELISA, and IHC with antibody or avidin/biotin probes (blocker is serum- and biotin-free)
• Short blocking times—less than 15 minutes for nitrocellulose or PVDF membranes; almost instantaneous for polystyrene microplate wells
Specifications
Blocking Buffer | |
StartingBlock™ | |
Upon receipt store at 4°C. |
Liquid | |
Single purified protein |