missing translation for 'onlineSavingsMsg'
Learn More

Shamrock Stationary Labeling Tape Dispenser

This Stationary Labeling Tape Dispenser accommodates multiple rolls of tape up to 6 in. wide.

Brand:  Shamrock SD-6

Product Code. 11847153

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • For use with 60 yard rolls of tape
  • Holds multiple rolls up to 6 in. wide
Specifications

Specifications

Stationary Tape Dispenser
Multiple 2160 in. rolls up to 6 in. wide
Specialty Tape and Label Rolls, 2160 in.
SD-6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers